ENA118036447 Enamine

4-tert-butyl-N-(3-chloropropanoyl)benzamide

  • MDL number MFCD09802096
  • Cas Number : 949393-09-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28904

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28904
Availability details for
Quantity
  • Availability
  • Add to Cart