ENA118036452 Enamine

5,6-difluoro-2-hydrazinyl-1H-1,3-benzodiazole

  • MDL number MFCD09802102
  • Cas Number : 175727-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28919
Availability details for
Quantity
  • Availability
  • Add to Cart