ENA118036454 Enamine

methyl 1-(aminosulfonyl)piperidine-4-carboxylate

  • MDL number MFCD09802104
  • Cas Number : 949444-70-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28923

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28923
Availability details for
Quantity
  • Availability
  • Add to Cart