ENA118036455 Enamine

4-fluoro-5-(1H-imidazol-1-yl)-2-nitroaniline

  • MDL number MFCD09802105
  • Cas Number : 216883-13-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28939

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28939
Availability details for
Quantity
  • Availability
  • Add to Cart