ENA118036486 Enamine

N-(4-bromo-2-chlorophenyl)-2-chloroacetamide

  • MDL number MFCD00157385
  • Cas Number : 195372-57-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24570

Safety Information

msds https://www.enaminestore.com/catalog/EN300-24570
Availability details for
Quantity
  • Availability
  • Add to Cart