ENA118036487 Enamine

2-chloro-N-[5-chloro-2-(4-ethoxyphenoxy)phenyl]acetamide

  • MDL number MFCD09383881
  • Cas Number : 930395-63-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24574

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24574
Availability details for
Quantity
  • Availability
  • Add to Cart