ENA118036488 Enamine

2-chloro-N-(4-difluoromethanesulfonylphenyl)acetamide

  • MDL number MFCD09383940
  • Cas Number : 930395-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24577

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24577
Availability details for
Quantity
  • Availability
  • Add to Cart