ENA118036489 Enamine

2-chloro-N-(2-difluoromethanesulfonylphenyl)acetamide

  • MDL number MFCD09376371
  • Cas Number : 930395-65-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24579

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24579
Availability details for
Quantity
  • Availability
  • Add to Cart