ENA118036491 Enamine

N-[3-(benzyloxy)propyl]-2-chloroacetamide

  • MDL number MFCD09388778
  • Cas Number : 13224-02-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24604

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24604
Availability details for
Quantity
  • Availability
  • Add to Cart