ENA118036495 Enamine

2-chloro-N-(3,5-difluorophenyl)acetamide

  • MDL number MFCD06409120
  • Cas Number : 874804-29-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-24613

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24613
Availability details for
Quantity
  • Availability
  • Add to Cart