ENA118036503 Enamine

2-chloro-N-[1-(1,1-dioxo-1lambda6-thiolan-3-yl)-3-methyl-1H-pyrazol-5-yl]acetamide

  • MDL number MFCD09383879
  • Cas Number : 956769-05-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-24675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-24675
Availability details for
Quantity
  • Availability
  • Add to Cart