ENA118036514 Enamine

3-chloro-N-(2-fluorophenyl)propanamide

  • MDL number MFCD01357336
  • Cas Number : 349097-66-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25822

Safety Information

msds https://www.enaminestore.com/catalog/EN300-25822
Availability details for
Quantity
  • Availability
  • Add to Cart