ENA118036521 Enamine

3-chloro-N-(2,3-dihydro-1H-inden-5-yl)propanamide

  • MDL number MFCD08593324
  • Cas Number : 908494-47-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-25847

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-25847
Availability details for
Quantity
  • Availability
  • Add to Cart