ENA118036528 Enamine

3,4-dihydro-2H-1,5-benzodioxepine-7-sulfonamide

  • MDL number MFCD07774225
  • Cas Number : 874842-53-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27012

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27012
Availability details for
Quantity
  • Availability
  • Add to Cart