ENA118036536 Enamine

2-{[(3-bromophenyl)methyl]amino}acetic acid

  • MDL number MFCD08443382
  • Cas Number : 501653-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27401

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27401
Availability details for
Quantity
  • Availability
  • Add to Cart