ENA118036541 Enamine

2-methyl-1-(4-pentylphenyl)propan-1-one

  • MDL number MFCD09373407
  • Cas Number : 116706-96-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-27408

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27408
Availability details for
Quantity
  • Availability
  • Add to Cart