ENA118036545 Enamine

4-(1H-imidazol-2-yl)phthalazin-1(2H)-one

  • MDL number MFCD00573033
  • Cas Number : 1955557-06-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27413

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27413
Availability details for
Quantity
  • Availability
  • Add to Cart