ENA118036589 Enamine

sodium [2-(3-phenoxyphenyl)-1,3-thiazol-4-yl]acetate

  • MDL number MFCD07288347
  • Cas Number : 1007016-49-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10194

Safety Information

msds https://www.enaminestore.com/catalog/EN300-10194
Availability details for
Quantity
  • Availability
  • Add to Cart