ENA118036591 Enamine

sodium 2-[2-(3-methoxyphenyl)-1,3-thiazol-4-yl]acetate

  • MDL number MFCD07288416
  • Cas Number : 1011426-50-8
  • Molecular Weight : 271.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 165-166
Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-10327

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-10327
Availability details for
Quantity
  • Availability
  • Add to Cart