ENA118036596 Enamine

methyl 4-(2-amino-1,3-thiazol-4-yl)-1H-pyrrole-2-carboxylate

  • MDL number MFCD09376370
  • Cas Number : 929974-98-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27481

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27481
Availability details for
Quantity
  • Availability
  • Add to Cart