ENA118036610 Enamine

4-(chloromethyl)-2-ethyl-1,3-thiazole

  • MDL number MFCD09040680
  • Cas Number : 40516-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27499

Safety Information

signalword Danger
msds https://www.enaminestore.com/catalog/EN300-27499
pictograms GHS05,GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart