ENA118036612 Enamine

[2-(4-methylphenyl)-1,3-thiazol-4-yl]acetonitrile

  • MDL number MFCD09376380
  • Cas Number : 16441-27-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27502

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27502
Availability details for
Quantity
  • Availability
  • Add to Cart