ENA118036622 Enamine

3-[2-(thiophen-2-yl)ethynyl]aniline

  • MDL number MFCD09376390
  • Cas Number : 930395-89-4
  • Molecular Weight : 199.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27575

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27575
Availability details for
Quantity
  • Availability
  • Add to Cart