ENA118036628 Enamine

4-methyl-2-(morpholin-4-yl)pentan-1-amine

  • MDL number MFCD08444595
  • Cas Number : 923249-21-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27583

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27583
Availability details for
Quantity
  • Availability
  • Add to Cart