ENA118036633 Enamine

5-hydroxy-5-(trifluoromethyl)-4,5-dihydro-1H-pyrazole-1-carbothioamide

  • MDL number MFCD03150934
  • Cas Number : 219986-56-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27589

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27589
Availability details for
Quantity
  • Availability
  • Add to Cart