ENA118036639 Enamine

3-[(oxolan-2-yl)methoxy]benzoic acid

  • MDL number MFCD06801287
  • Cas Number : 883536-39-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27597

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27597
Availability details for
Quantity
  • Availability
  • Add to Cart