ENA118036640 Enamine

4-(4-methylphenoxy)phenol

  • MDL number MFCD08446909
  • Cas Number : 35094-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27599

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27599
Availability details for
Quantity
  • Availability
  • Add to Cart