ENA118036648 Enamine

4-(trifluoromethyl)quinoline-3-carboxylic acid

  • MDL number MFCD08458130
  • Cas Number : 155495-82-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27608

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27608
Availability details for
Quantity
  • Availability
  • Add to Cart