ENA118036676 Enamine

1-(2-fluorophenyl)-5-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD04122761
  • Cas Number : 618092-40-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27674

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27674
Availability details for
Quantity
  • Availability
  • Add to Cart