ENA118036682 Enamine

5,7-dimethoxycinnoline-3-carboxylic acid

  • MDL number MFCD09383937
  • Cas Number : 929975-20-2
  • Molecular Weight : 234.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-27682

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27682
Availability details for
Quantity
  • Availability
  • Add to Cart