ENA118036689 Enamine

1-[2-(2-furyl)-4-hydroxy-6-methylpyrimidin-5-yl]ethanone

  • MDL number MFCD09702176
  • Cas Number : 930396-01-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27697

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27697
Availability details for
Quantity
  • Availability
  • Add to Cart