ENA118036691 Enamine

methyl 3-[2-(3-aminophenyl)ethynyl]benzoate

  • MDL number MFCD09388769
  • Cas Number : 930396-02-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-27699

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27699
Availability details for
Quantity
  • Availability
  • Add to Cart