ENA118036699 Enamine

8-chloro-5-nitroquinoline

  • MDL number MFCD00466079
  • Cas Number : 22539-55-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 144-146
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27726

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-27726
Availability details for
Quantity
  • Availability
  • Add to Cart