ENA118036705 Enamine

2-benzyl-9-methyl-2,3-dihydro-1H-pyrrolo[3,4-b]quinolin-1-one

  • MDL number MFCD09396705
  • Cas Number : 941083-73-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-27869

Safety Information

msds https://www.enaminestore.com/catalog/EN300-27869
Availability details for
Quantity
  • Availability
  • Add to Cart