ENA118036708 Enamine

1-methyl-5-(1H-pyrrol-1-yl)-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD01871363
  • Cas Number : 293758-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28232

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28232
Availability details for
Quantity
  • Availability
  • Add to Cart