ENA118036715 Enamine

5-[(3-chloro-4-fluorophenyl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD06496550
  • Cas Number : 893724-17-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28585

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28585
Availability details for
Quantity
  • Availability
  • Add to Cart