ENA118036716 Enamine

5-[(3-chloro-4-methylphenyl)methyl]-2,3-dihydro-1,3-thiazol-2-imine

  • MDL number MFCD26143609
  • Cas Number : 501349-36-6
  • Molecular Weight : 238.74

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28587

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28587
Availability details for
Quantity
  • Availability
  • Add to Cart