ENA118036717 Enamine

5-[(naphthalen-1-yl)methyl]-1,3-thiazol-2-amine

  • MDL number MFCD00547830
  • Cas Number : 207463-35-2
  • Molecular Weight : 240.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 180-181
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28589

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P363,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-28589
pictograms GHS07
hstatements H302,H312,H315,H319,H332,H335
Availability details for
Quantity
  • Availability
  • Add to Cart