ENA118036732 Enamine

5-chloroquinolin-8-amine

  • MDL number MFCD00461243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-28776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28776
Availability details for
Quantity
  • Availability
  • Add to Cart