ENA118036733 Enamine

N-(5-amino-2-fluorophenyl)acetamide

  • MDL number MFCD09049668
  • Cas Number : 75001-47-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28777

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28777
Availability details for
Quantity
  • Availability
  • Add to Cart