ENA118036740 Enamine

5-[(4-methoxyphenyl)methyl]-2,3-dihydro-1,3-thiazol-2-imine

  • MDL number MFCD23145310
  • Cas Number : 299953-06-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28789

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28789
Availability details for
Quantity
  • Availability
  • Add to Cart