ENA118036750 Enamine

sodium 2-{8-oxatricyclo[7.4.0.0,2,7]trideca-1(9),2(7),3,5,10,12-hexaen-4-yloxy}acetate

  • MDL number MFCD07287069
  • Cas Number : 1049761-58-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 220-221
Purity 95
Salt Data Na+
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-11953

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-11953
Availability details for
Quantity
  • Availability
  • Add to Cart