ENA118036758 Enamine

N,N,N-trimethyl(1-methyl-1H-pyrrol-2-yl)methanaminium iodide

  • MDL number MFCD07310901
  • Cas Number : 54828-80-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 16...18
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-13697

Safety Information

msds https://www.enaminestore.com/catalog/EN300-13697
Availability details for
Quantity
  • Availability
  • Add to Cart