ENA118036760 Enamine

1-(ethylsulfonyl)piperidin-4-amine

  • MDL number MFCD06805737
  • Cas Number : 759456-76-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 51...53
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28817

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28817
Availability details for
Quantity
  • Availability
  • Add to Cart