ENA118036767 Enamine

N-(5-amino-2-fluorophenyl)methanesulfonamide

  • MDL number MFCD09050848
  • Cas Number : 123343-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28826

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28826
Availability details for
Quantity
  • Availability
  • Add to Cart