ENA118036776 Enamine

N,3-diphenyl-1,2,4-thiadiazol-5-amine

  • MDL number MFCD00159801
  • Cas Number : 17467-60-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28891

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28891
Availability details for
Quantity
  • Availability
  • Add to Cart