ENA118036791 Enamine

3-Benzyl-7-(3-methyl-butyl)-3,7-dihydro-purine-2,6-dione

  • MDL number MFCD06671612
  • Cas Number : 1052541-60-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 108...110
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-05295

Safety Information

msds https://www.enaminestore.com/catalog/EN300-05295
Availability details for
Quantity
  • Availability
  • Add to Cart