ENA118036801 Enamine

4-(benzoylamino)benzoic acid

  • MDL number MFCD00451153
  • Cas Number : 582-80-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 297...299
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28937

Safety Information

msds https://www.enaminestore.com/catalog/EN300-28937
Availability details for
Quantity
  • Availability
  • Add to Cart