ENA118036804 Enamine

1-(2-methylpropanoyl)piperidine-3-carboxylic acid

  • MDL number MFCD09050645
  • Cas Number : 926214-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-28945

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-28945
Availability details for
Quantity
  • Availability
  • Add to Cart