ENA118036824 Enamine

4-[(2,4-difluorobenzoyl)amino]butanoic acid

  • MDL number MFCD09388779
  • Cas Number : 930395-60-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-18568

Safety Information

msds https://www.enaminestore.com/catalog/EN300-18568
Availability details for
Quantity
  • Availability
  • Add to Cart